tert-Butyl N-(azetidin-3-yl)-N-methylcarbamate hydrochloride

Lieferant: Apollo Scientific
Achtung

OR308031-1G OR308031-25G OR308031-5G
APOSOR308031-1GEA 0 EUR
APOSOR308031-1G APOSOR308031-25G APOSOR308031-5G
tert-Butyl N-(azetidin-3-yl)-N-methylcarbamate hydrochloride
tert-Butyl N-(azetidin-3-yl)-N-methylcarbamate hydrochloride
Formel: C₉H₁₉ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12963843
CAS-Nummer: 943060-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR