3-Boc-aminoethylazetidine

Lieferant: Apollo Scientific

Synonyme: 3-[2-(Boc-amino)ethyl]azetidine, 3-(N-BOC)-(2-aminoethyl)azetidine, 3-Azetidinecarboxylic acid, 1-(2-aminoethyl)-, 1,1-dimethylethyl ester, 3-Boc-Aminoethylazetidine, 2-Methyl-2-propanyl 1-(2-aminoethyl)-3-azetidinecarboxylate

OR308033-1G
APOSOR308033-1GEA 980 EUR
APOSOR308033-1G
3-Boc-aminoethylazetidine
3-Boc-aminoethylazetidine
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,282 g/mol
Schmelzpunkt: 84…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09878605
CAS-Nummer: 162696-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR