5-Chlor-1H-pyrrolo[2,3-b]pyridin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Chloro-7-azaindole
OR308035-25G
APOSOR308035-25GEA
77
EUR
APOSOR308035-25G
5-Chlor-1H-pyrrolo[2,3-b]pyridin
5-Chloro-1H-pyrrolo[2,3-b]pyridine
Formel:
C₇H₅ClN₂ Molecular Weight: 152,58 g/mol Schmelzpunkt: 161…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659676 CAS-Nummer: 866546-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...