1H-Pyrrolo[2,3-b]pyridin-4-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-1H-pyrrolo[2,3-b]pyridine

OR308038-1G
APOSOR308038-1GEA 155 EUR
APOSOR308038-1G
1H-Pyrrolo[2,3-b]pyridin-4-amine
1H-Pyrrolo[2,3-b]pyridin-4-amine
Formel: C₇H₇N₃
Molecular Weight: 133,15 g/mol
Schmelzpunkt: 122…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07778366
CAS-Nummer: 74420-00-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR