1H-Pyrrolo[2,3-b]pyridine-5-carbaldehyde

Lieferant: Apollo Scientific

OR308040-1G
APOSOR308040-1GEA 89.9 EUR
APOSOR308040-1G
1H-Pyrrolo[2,3-b]pyridine-5-carbaldehyde
1H-Pyrrolo[2,3-b]pyridine-5-carbaldehyde
Formel: C₈H₆N₂O
Molecular Weight: 146.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08272241
CAS-Nummer: 849067-90-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR