4-Chloro-2-iodo-7-azaindole
Lieferant: Apollo Scientific
Synonyme:
4-Chloro-2-iodo-1H-pyrrolo[2,3-b]pyridine
OR308046-1G
APOSOR308046-1GEA
220
EUR
APOSOR308046-1G
4-Chloro-2-iodo-7-azaindole
4-Chloro-2-iodo-7-azaindole
Formel:
C₇H₄ClIN₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
940948-29-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...