1-(Benzenesulphonyl)-4-chloro-2-iodopyrrolo[2,3-b]pyridine

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-iodo-1-(phenylsulphonyl)-1H-pyrrolo[2,3-b]pyridine

OR308048-1G
APOSOR308048-1GEA 214 EUR
APOSOR308048-1G
1-(Benzenesulphonyl)-4-chloro-2-iodopyrrolo[2,3-b]pyridine
1-(Benzenesulphonyl)-4-chloro-2-iodopyrrolo[2,3-b]pyridine
Formel: C₁₃H₈ClIN₂O₂S
Molecular Weight: 418,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12923119
CAS-Nummer: 940948-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR