2-Methylindazole-7-boronic acid pinacol ester

Lieferant: Apollo Scientific

OR308059-1G OR308059-5G
APOSOR308059-1GEA 255 EUR
APOSOR308059-1G APOSOR308059-5G
2-Methylindazole-7-boronic acid pinacol ester
2-Methylindazole-7-boronic acid pinacol ester
Formel: C₁₄H₁₉BN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11109457
CAS-Nummer: 845751-67-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR