1-Boc-piperazin-2-carbonsäure

Lieferant: Apollo Scientific

Synonyme: 1-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid

OR308063-5G
APOSOR308063-5GEA 92.8 EUR
APOSOR308063-5G
1-Boc-piperazin-2-carbonsäure
1-Boc-piperazin-2-carbonsäure
Formel: C₁₀H₁₈N₂O₄
Molecular Weight: 230,2600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02682403
CAS-Nummer: 1214196-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR