(S)-1-tert-Butyl-2-methylpiperazine-1,2-dicarboxylate

Lieferant: Apollo Scientific
Achtung

OR308067-1G OR308067-5G
APOSOR308067-1GEA 97.3 EUR
APOSOR308067-1G APOSOR308067-5G
(S)-1-tert-Butyl-2-methylpiperazine-1,2-dicarboxylate
(S)-1-tert-Butyl-2-methylpiperazine-1,2-dicarboxylate
Formel: C₁₁H₂₀N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD04115328
CAS-Nummer: 796096-64-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR