(4-Methyl-2-phenyl-1,3-oxazol-5-yl)methylamine 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
[5-(Aminomethyl)-4-methyl-1,3-oxazol-2-yl]benzene, 5-(Aminomethyl)-4-methyl-2-phenyl-1,3-oxazole
OR30807-250MG
APOSOR30807-250MGEA
790
EUR
APOSOR30807-250MG
(4-Methyl-2-phenyl-1,3-oxazol-5-yl)methylamine 95%
(4-Methyl-2-phenyl-1,3-oxazol-5-yl)methylamine
Formel:
C₁₁H₁₂N₂O Molecular Weight: 188.226 g/mol Schmelzpunkt: 47…49 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11109320 CAS-Nummer: 165735-97-7 UN: 3259 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...