(R)-1-tert-Butyl-3-methyl piperazine-1,3-dicarboxylate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate

OR308070-1G
APOSOR308070-1GEA 33.5 EUR
APOSOR308070-1G
(R)-1-tert-Butyl-3-methyl piperazine-1,3-dicarboxylate 95%
(R)-1-tert-Butyl-3-methyl piperazine-1,3-dicarboxylate
Formel: C₁₁H₂₀N₂O₄
Molecular Weight: 244,2875 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04115325
CAS-Nummer: 438631-77-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR