(R)-1-Boc-3-(hydroxymethyl)piperazin 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-tert-Butyl 3-(hydroxymethyl)piperazine-1-carboxylate, (3R)-3-(Hydroxymethyl)piperazine, N1-BOC protected

OR308071-1G
APOSOR308071-1GEA 21.9 EUR
APOSOR308071-1G
(R)-1-Boc-3-(hydroxymethyl)piperazin 99%
(R)-1-Boc-3-(hydroxymethyl)piperazin
Formel: C₁₀H₂₀N₂O₃
Molecular Weight: 216,2774 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863888
CAS-Nummer: 278788-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR