Pyrazolo[1,5-a]pyridin-3-carbonsäure 95%

Lieferant: Apollo Scientific
Achtung

OR308077-5G
APOSOR308077-5GEA 140 EUR
APOSOR308077-5G
Pyrazolo[1,5-a]pyridin-3-carbonsäure 95%
Pyrazolo[1,5-a]pyridin-3-carbonsäure
Formel: C₈H₆N₂O₂
Molecular Weight: 162.1454 g/mol
Schmelzpunkt: 199…201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00229214
CAS-Nummer: 16205-46-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR