1H-Pyrazolo[3,4-b]pyridine-5-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR308078-1G
APOSOR308078-1GEA 121 EUR
APOSOR308078-1G
1H-Pyrazolo[3,4-b]pyridine-5-carboxylic acid
1H-Pyrazolo[3,4-b]pyridine-5-carboxylic acid
Formel: C₇H₅N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09832891
CAS-Nummer: 952182-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR