[1,2,4]Triazolo[4,3-a]pyridine-7-carboxylic acid

Lieferant: Apollo Scientific

OR308083-1G
APOSOR308083-1GEA 0 EUR
APOSOR308083-1G
[1,2,4]Triazolo[4,3-a]pyridine-7-carboxylic acid
[1,2,4]Triazolo[4,3-a]pyridine-7-carboxylic acid
Formel: C₇H₅N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD17015978
CAS-Nummer: 1234616-66-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR