6-Quinazolinecarboxylic acid

Lieferant: Apollo Scientific

OR308084-100MG OR308084-250MG
APOSOR308084-100MGEA 0 EUR
APOSOR308084-100MG APOSOR308084-250MG
6-Quinazolinecarboxylic acid
6-Quinazolinecarboxylic acid
Formel: C₉H₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11520386
CAS-Nummer: 676326-53-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR