cis-3-(Boc-amino)cyclobutanemethanol
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl [cis-3-(hydroxymethyl)cyclobutyl]carbamate, tert-Butyl (cis-3-(hydroxymethyl)cyclobutyl)carbamate, tert-butyl N-[cis-3-(hydroxymethyl)cyclobutyl]carbamate
OR308096-1G
APOSOR308096-1GEA
125
EUR
APOSOR308096-1G
cis-3-(Boc-amino)cyclobutanemethanol
cis-3-(Boc-amino)cyclobutanemethanol
Formel:
C₁₀H₁₉NO₃ Molecular Weight: 201,266 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08436074 CAS-Nummer: 142733-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...