trans-3-Aminocyclobutanol hydrochloride

Lieferant: Apollo Scientific
Achtung

OR308098-1G
APOSOR308098-1GEA 360 EUR
APOSOR308098-1G
trans-3-Aminocyclobutanol hydrochloride
trans-3-Aminocyclobutanol hydrochloride
Formel: C₄H₁₀ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD17214232
CAS-Nummer: 1205037-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR