3-Benzyloxycyclobutanecarboxylic acid

Lieferant: Apollo Scientific

Synonyme: 3-(benzyloxy)cyclobutane-1-carboxylic acid, 3-(Benzyloxy)cyclobutanecarboxylic acid

OR308099-1G
APOSOR308099-1GEA 65.4 EUR
APOSOR308099-1G
3-Benzyloxycyclobutanecarboxylic acid
3-Benzyloxycyclobutanecarboxylic acid
Formel: C₁₂H₁₄O₃
Molecular Weight: 206,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00512751
CAS-Nummer: 4958-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR