2-(Piperidin-1-yl)-1,3-thiazole-5-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR30810-250MG
APOSOR30810-250MGEA 337 EUR
APOSOR30810-250MG
2-(Piperidin-1-yl)-1,3-thiazole-5-carboxylic acid
2-(Piperidin-1-yl)-1,3-thiazole-5-carboxylic acid
Formel: C₉H₁₂N₂O₂S
Molecular Weight: 212,269 g/mol
Schmelzpunkt: 146…151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11506352
CAS-Nummer: 180403-13-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR