tert-Butyl-6-oxo-2-azaspiro[3.3]heptane-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 6-oxo-2-azaspiro[3.3]heptane-2-carboxylate
OR308105-250MG
APOSOR308105-250MGEA
21.9
EUR
APOSOR308105-250MG
tert-Butyl-6-oxo-2-azaspiro[3.3]heptane-2-carboxylate
tert-Butyl-6-oxo-2-azaspiro[3.3]heptane-2-carboxylate
Formel:
C₁₁H₁₇NO₃ Molecular Weight: 211,2576 g/mol Siedepunkt: 314,7 °C Flammpunkt: 114,1 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD15071430 CAS-Nummer: 1181816-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...