2,7-Diazaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 2,7-diazaspiro[3.5]nonane-7-carboxylate

OR308112-1G
APOSOR308112-1GEA 49.3 EUR
APOSOR308112-1G
2,7-Diazaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester 98%
2,7-Diazaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester
Formel: C₁₂H₂₂N₂O₂
Molecular Weight: 226,32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09839640
CAS-Nummer: 896464-16-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR