2,7-Diazaspiro[3.5]nonane-2-carboxylic acid tert-butyl ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-butyl 2,7-diazaspiro[3.5]nonane-2-carboxylate, 2-(tert-Butoxycarbonyl)-2,7-diazaspiro[3.5]nonane

OR308113-5G
APOSOR308113-5GEA 68.3 EUR
APOSOR308113-5G
2,7-Diazaspiro[3.5]nonane-2-carboxylic acid tert-butyl ester 97%
2,7-Diazaspiro[3.5]nonane-2-carboxylic acid tert-butyl ester
Formel: C₁₂H₂₂N₂O₂
Molecular Weight: 226.32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09834803
CAS-Nummer: 236406-55-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR