N-(2-Aminospiro[3.3]hept-6-yl)carbamic acid-tert-butyl ester
Lieferant: Apollo Scientific
|
Achtung
|
OR308114-1G
APOSOR308114-1GEA
0
EUR
APOSOR308114-1G
N-(2-Aminospiro[3.3]hept-6-yl)carbamic acid-tert-butyl ester
N-(2-Aminospiro[3.3]hept-6-yl)carbamic acid-tert-butyl ester
Formel:
C₁₂H₂₂N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09054707 CAS-Nummer: 1239589-52-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...