(6-Oxospiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester

Lieferant: Apollo Scientific

Synonyme: tert-Butyl (6-oxospiro[3.3]heptan-2-yl)carbamate, tert-butyl N-{6-oxospiro[3.3]heptan-2-yl}carbamate

OR308116-1G
APOSOR308116-1GEA 288 EUR
APOSOR308116-1G
(6-Oxospiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester
(6-Oxospiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester
Formel: C₁₂H₁₉NO₃
Molecular Weight: 225,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11858160
CAS-Nummer: 1118786-86-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR