tert-Butyl-3-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 3-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate

OR308119-1G
APOSOR308119-1GEA 58 EUR
APOSOR308119-1G
tert-Butyl-3-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate
tert-Butyl-3-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate
Formel: C₁₃H₂₂N₂O₃
Molecular Weight: 254,33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11111226
CAS-Nummer: 169206-67-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR