8-Boc-2,8-Diazaspiro[4.5]decane

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 2,8-diazaspiro[4.5]decane-8-carboxylate, 2,8-Diazaspiro[4.5]decane-8-carboxylic acid-tert-butyl ester

OR308120-250MG
APOSOR308120-250MGEA 104 EUR
APOSOR308120-250MG
8-Boc-2,8-Diazaspiro[4.5]decane
8-Boc-2,8-Diazaspiro[4.5]decane
Formel: C₁₃H₂₄N₂O₂
Molecular Weight: 240,342 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09608078
CAS-Nummer: 236406-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR