4-Methylpiperidin-4-ol

Lieferant: Apollo Scientific
Gefahr

OR308124-5G
APOSOR308124-5GEA 82.8 EUR
APOSOR308124-5G
4-Methylpiperidin-4-ol
4-Methylpiperidin-4-ol
Formel: C₆H₁₃NO
Molecular Weight: 115.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100993
CAS-Nummer: 3970-68-1
UN: 3263
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR