(3S)-3-Amino-N-methylpyrrolidine, N-BOC protected

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl methyl[(3S)-pyrrolidin-3-yl]carbamate, (S)-tert-Butyl methyl(pyrrolidin-3-yl)carbamate

OR308137-1G
APOSOR308137-1GEA 127 EUR
APOSOR308137-1G
(3S)-3-Amino-N-methylpyrrolidine, N-BOC protected
(3S)-3-Amino-N-methylpyrrolidine, N-BOC protected
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,278 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08703388
CAS-Nummer: 169750-01-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR