(R)-4-(tert-Butoxycarbonyl)morpholine-2-carboxylic acid

Lieferant: Apollo Scientific

OR308151-5G
APOSOR308151-5GEA 111 EUR
APOSOR308151-5G
(R)-4-(tert-Butoxycarbonyl)morpholine-2-carboxylic acid
(R)-4-(tert-Butoxycarbonyl)morpholine-2-carboxylic acid
Formel: C₁₀H₁₇NO₅
Molecular Weight: 231,2457 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09260604
CAS-Nummer: 884512-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR