(S)-4-Boc-morpholine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR308152-5G
APOSOR308152-5GEA 137 EUR
APOSOR308152-5G
(S)-4-Boc-morpholine-2-carboxylic acid
(S)-4-Boc-morpholine-2-carboxylic acid
Formel: C₁₀H₁₇NO₅
Schmelzpunkt: 127…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06200662
CAS-Nummer: 868689-63-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR