(S)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR308160-1G
APOSOR308160-1GEA 87 EUR
APOSOR308160-1G
(S)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid
(S)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid
Formel: C₁₀H₁₇NO₅
Molecular Weight: 231,2457 g/mol
Schmelzpunkt: 169…173 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114904
CAS-Nummer: 783350-37-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR