(R)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(R)-4-Boc-morpholine-3-carboxylic acid
OR308161-5G
APOSOR308161-5GEA
209
EUR
APOSOR308161-5G
(R)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid 95%
(R)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid
Formel:
C₁₀H₁₇NO₅ Molecular Weight: 231,25 g/mol Schmelzpunkt: 181 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD04114909 CAS-Nummer: 869681-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...