Methyl (3R)-morpholine-3-carboxylate hydrochloride

Lieferant: Apollo Scientific

Synonyme: (R)-Methyl morpholine-3-carboxylate hydrochloride

OR308166-1G
APOSOR308166-1GEA 172 EUR
APOSOR308166-1G
Methyl (3R)-morpholine-3-carboxylate hydrochloride
Methyl (3R)-morpholine-3-carboxylate hydrochloride
Formel: C₆H₁₁NO₃·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD09910332
CAS-Nummer: 1187929-55-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR