(S)-Methyl morpholine-3-carboxylate hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR308167-1G
APOSOR308167-1GEA
127
EUR
APOSOR308167-1G
(S)-Methyl morpholine-3-carboxylate hydrochloride
(S)-Methyl morpholine-3-carboxylate hydrochloride
Formel:
C₆H₁₂ClNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09910333 CAS-Nummer: 1447972-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...