Methyl morpholine-2-carboxylate hydrochloride

Lieferant: Apollo Scientific

OR308170-5G
APOSOR308170-5GEA 63.8 EUR
APOSOR308170-5G
Methyl morpholine-2-carboxylate hydrochloride
Methyl morpholine-2-carboxylate hydrochloride
Formel: C₆H₁₂ClNO₃
Molecular Weight: 181.62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17167182
CAS-Nummer: 937063-34-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR