Methyl (2R)-morpholine-2-carboxylate hydrochloride

Lieferant: Apollo Scientific

Synonyme: (R)-Methyl morpholine-2-carboxylate hydrochloride

OR308171-1G
APOSOR308171-1GEA 77 EUR
APOSOR308171-1G
Methyl (2R)-morpholine-2-carboxylate hydrochloride
Methyl (2R)-morpholine-2-carboxylate hydrochloride
Formel: C₆H₁₁NO₃·ClH
Molecular Weight: 181,617 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21606371
CAS-Nummer: 1352709-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR