(S)-3-Isopropylmorpholine

Lieferant: Apollo Scientific

OR308182-1G
APOSOR308182-1GEA 203 EUR
APOSOR308182-1G
(S)-3-Isopropylmorpholine
(S)-3-Isopropylmorpholine
Formel: C₇H₁₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD11519112
CAS-Nummer: 77897-21-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR