(2S)-2-Benzylmorpholine

Lieferant: Apollo Scientific

OR308190-250MG
APOSOR308190-250MGEA 337 EUR
APOSOR308190-250MG
(2S)-2-Benzylmorpholine
(2S)-2-Benzylmorpholine
Formel: C₁₁H₁₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD14635496
CAS-Nummer: 947687-18-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR