Boc-(4-carboxymethyl)piperazine
Lieferant: Apollo Scientific
|
Achtung
|
OR30823-25G
APOSOR30823-25GEA
154
EUR
APOSOR30823-25G
Boc-(4-carboxymethyl)piperazine
Boc-(4-carboxymethyl)piperazine
Formel:
C₁₁H₂₀N₂O₄ Schmelzpunkt: 186,5…189.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02682402 CAS-Nummer: 156478-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...