5-Bromo-4-methoxy-1H-indazole

Lieferant: Apollo Scientific
Achtung

OR30831-250MG
APOSOR30831-250MGEA 77 EUR
APOSOR30831-250MG
5-Bromo-4-methoxy-1H-indazole
5-Bromo-4-methoxy-1H-indazole
Formel: C₈H₇BrN₂O
Molecular Weight: 227,058 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09261245
CAS-Nummer: 850363-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR