6-(Aminomethyl)-1H-indazole

Lieferant: Apollo Scientific
Achtung

OR30834-250MG
APOSOR30834-250MGEA 288 EUR
APOSOR30834-250MG
6-(Aminomethyl)-1H-indazole
6-(Aminomethyl)-1H-indazole
Formel: C₈H₉N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09475774
CAS-Nummer: 710943-26-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR