5-Bromo-6-methoxy-1H-indazole
Lieferant: Apollo Scientific
|
Achtung
|
OR30839-1G
APOSOR30839-1GEA
40.6
EUR
APOSOR30839-1G
5-Bromo-6-methoxy-1H-indazole
5-Bromo-6-methoxy-1H-indazole
Formel:
C₈H₇BrN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD11007880 CAS-Nummer: 152626-78-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...