3-Methyl-1H-indazol-6-boronsäure 95+%

Lieferant: Apollo Scientific

Synonyme: 3-Methyl-1H-indazole-6-boronic Acid, 3-Methylindazole-6-boronic acid

OR30840-250MG
APOSOR30840-250MGEA 89.9 EUR
APOSOR30840-250MG
3-Methyl-1H-indazol-6-boronsäure 95+%
3-Methyl-1H-indazole-6-boronic acid
Formel: C₈H₉BN₂O₂
Molecular Weight: 175,9800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044596
CAS-Nummer: 1245816-26-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR