6-Hydroxy-1-methyl-1H-indazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methyl-1H-indazol-6-ol

OR30844-250MG
APOSOR30844-250MGEA 82.8 EUR
APOSOR30844-250MG
6-Hydroxy-1-methyl-1H-indazole
6-Hydroxy-1-methyl-1H-indazole
Formel: C₈H₈N₂O
Molecular Weight: 148,162 g/mol
Schmelzpunkt: 215…222 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00456074
CAS-Nummer: 118933-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR