1-Methyl-1H-indazole-6-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Cyano-1-methyl-1H-indazole

OR30852-250MG
APOSOR30852-250MGEA 78.3 EUR
APOSOR30852-250MG
1-Methyl-1H-indazole-6-carbonitrile
1-Methyl-1H-indazole-6-carbonitrile
Formel: C₉H₇N₃
Molecular Weight: 157,172 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12028576
CAS-Nummer: 267413-29-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR