4-(Aminomethyl)-1H-indazole
Lieferant: Apollo Scientific
|
Achtung
|
OR30869-250MG
APOSOR30869-250MGEA
134
EUR
APOSOR30869-250MG
4-(Aminomethyl)-1H-indazole
4-(Aminomethyl)-1H-indazole
Formel:
C₈H₉N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD10696781 CAS-Nummer: 944898-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...