N,N-Diethyl-4-nitrosoanilin 98%

Lieferant: Apollo Scientific

Synonyme: 4-Nitroso-N,N-diethylaniline

OR30878-25G
APOSOR30878-25GEA 299 EUR
APOSOR30878-25G
N,N-Diethyl-4-nitrosoanilin 98%
N,N-Diethyl-4-nitrosoanilin
Formel: C₁₀H₁₄N₂O
Molecular Weight: 178,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002064
CAS-Nummer: 120-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR