4-Acetyl-2-methyl-2H-indazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Ethanoyl-2-methyl-2H-indazole, 1-(2-Methyl-2H-indazol-4-yl)ethan-1-one

OR30892-500MG
APOSOR30892-500MGEA 381 EUR
APOSOR30892-500MG
4-Acetyl-2-methyl-2H-indazole
4-Acetyl-2-methyl-2H-indazole
Formel: C₁₀H₁₀N₂O
Molecular Weight: 174,199 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11869770
CAS-Nummer: 1159511-27-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR